4-Amino-2,2,6,6-tetramethylpiperidinooxy, 97%, free radical
4-Amino-2,2,6,6-tetramethylpiperidinooxy, 97%, free radical
4-Amino-2,2,6,6-tetramethylpiperidinooxy, 97%, free radical
Thermo Scientific Chemicals

4-Amino-2,2,6,6-tetramethylpiperidinooxy, 97%, free radical

CAS: 14691-88-4 | C9H19N2O | 171.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 172730010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS14691-88-4
IUPAC Name1-hydroxy-2,2,6,6-tetramethylpiperidin-4-aminium
Molecular FormulaC9H21N2O
InChI KeyAYSOUYKQIUYGFR-UHFFFAOYSA-O
SMILESCC1(C)CC([NH3+])CC(C)(C)N1O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Form)Adhering crystals or needles or moist mass
Appearance (Color)Red to red-brown
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Polymerization reagent
RUO – Research Use Only