Product Image
Applied Biosystems™

Seal, Isolation, Rheodyne Valve (for 140C instrument)

Have Questions?
Catalog NumberQuantity
1730014
also known as 173-0014
1 Unit
Catalog number 1730014
also known as 173-0014
Price (BRL)
-
Quantity:
1 Unit
This is a replacement part for the 140C Instrument.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Procise Protein Sequencer
Quantity1 Unit
Shipping ConditionRoom Temperature
Product TypeProtein Sequencer Seal
Unit SizeEach
Contents & Storage
Store at room temperature.