2-Hydroxyisobutyric acid, 98%
2-Hydroxyisobutyric acid, 98%
2-Hydroxyisobutyric acid, 98%
Thermo Scientific Chemicals

2-Hydroxyisobutyric acid, 98%

CAS: 594-61-6 | C4H8O3 | 104.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
1 kg
Packaging:
Plastic bottle
Catalog number 173101000
Price (USD)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS594-61-6
IUPAC Name2-hydroxy-2-methylpropanoic acid
Molecular FormulaC4H8O3
InChI KeyBWLBGMIXKSTLSX-UHFFFAOYSA-N
SMILESCC(C)(O)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Melting point75°C to 81°C
Appearance (Form)Crystalline powder or crystals or needles
Infrared spectrumConforms
Titration with NaOH>=97.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only