Sodium methoxide, pure, anhydrous powder
Sodium methoxide, pure, anhydrous powder
Sodium methoxide, pure, anhydrous powder
Thermo Scientific Chemicals

Sodium methoxide, pure, anhydrous powder

CAS: 124-41-4 | CH3NaO | 54.02 g/mol
数量:
5 g
100 g
500 g
2.5 kg
パッケージ:
すべて表示
Glass bottle
製品番号(カタログ番号) 173120050
価格(JPY)
-
数量:
5 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS124-41-4
IUPAC Namesodium methanolate
Molecular FormulaCH3NaO
InChI KeyWQDUMFSSJAZKTM-UHFFFAOYSA-N
SMILES[Na+].C[O-]
さらに表示
Appearance (Color)White to yellow
NaOH + Na2CO3 (as NaOH)=<2 %
Appearance (Form)Powder
Alkalinity>=99 % (Total)
Titration>=97 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only