Thermo Scientific™
Nunc™ Cell Factory™ Systems with Active Gassing
Create an ideal environment for culturing oxygen-consuming and pH sensitive cells using Thermo Scientific™ Nunc™ Cell Factory™ Systems with Active Gassing. Flow system helps ensure a controlled, contamination-free atmosphere in each culture tray by equally distributing the user-specified gas mixture. Available in 4-, 10- and 40-tray models.
Back to top
  • Achieve greater yields with controlled cell culture environments equally distributed within each layer
  • Optimize production capacity without changing cell culture platforms
  • Develop industrial scale solutions with the utilization of an active gassing Cell Factory™ Incubator