Fluorescein, disodium salt, pure
Fluorescein, disodium salt, pure
Fluorescein, disodium salt, pure
Fluorescein, disodium salt, pure
Thermo Scientific Chemicals

Fluorescein, disodium salt, pure

CAS: 518-47-8 | C20H10Na2O5 | 376.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Glass Bottle
Catalog number 173241000
Price (EUR)
58,00
Each
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
58,00
Each
Add to cart
Chemical Identifiers
CAS518-47-8
IUPAC Namedisodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Molecular FormulaC20H10Na2O5
InChI KeyNJDNXYGOVLYJHP-UHFFFAOYSA-L
SMILES[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
UV Lambda max488 to 491 nm (in buffer pH 8)
Appearance (Color)Orange-red to green-brown to dark brown
Infrared spectrumConforms
Loss on drying=<10 % (105°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only