1,3-Dichloroacetone, 96%
1,3-Dichloroacetone, 96%
1,3-Dichloroacetone, 96%
Thermo Scientific Chemicals

1,3-Dichloroacetone, 96%

CAS: 534-07-6 | C3H4Cl2O | 126.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 173632500
Price (EUR)
245,00
Each
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
245,00
Each
Chemical Identifiers
CAS534-07-6
IUPAC Name1,3-dichloropropan-2-one
Molecular FormulaC3H4Cl2O
InChI KeySUNMBRGCANLOEG-UHFFFAOYSA-N
SMILESClCC(=O)CCl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Infrared spectrumConforms
Melting point38°C to 44°C
Appearance (Form)Crystalline powder or crystals and/or chunks
GC>=95 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only