Undecanoic acid, 99%
Undecanoic acid, 99%
Undecanoic acid, 99%
Thermo Scientific Chemicals

Undecanoic acid, 99%

CAS: 112-37-8 | C11H22O2 | 186.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 173970250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS112-37-8
IUPAC Nameundecanoic acid
Molecular FormulaC11H22O2
InChI KeyZDPHROOEEOARMN-UHFFFAOYSA-N
SMILESCCCCCCCCCCC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=98.5 %
Infrared spectrumConforms
Appearance (Color)White to light yellow
GC>=98.5 %
Appearance (Form)Low melting solid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only