1-Iodopropane, 99%, stabilized
1-Iodopropane, 99%, stabilized
1-Iodopropane, 99%, stabilized
Thermo Scientific Chemicals

1-Iodopropane, 99%, stabilized

CAS: 107-08-4 | C3H7I | 169.99 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1740500101 LGlass bottle
Catalog number 174050010
Price (USD)
-
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS107-08-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=98.5 %
Refractive index1.5030 to 1.5060 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only