2-Hydroxyethyl salicylate, 98+%, Thermo Scientific Chemicals
2-Hydroxyethyl salicylate, 98+%, Thermo Scientific Chemicals
2-Hydroxyethyl salicylate, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Hydroxyethyl salicylate, 98+%, Thermo Scientific Chemicals

製品番号(カタログ番号) 174100010
価格(JPY)
-
一括またはカスタム形式をリクエストする
化学物質識別子
CAS87-28-5
IUPAC Name2-hydroxyethyl 2-hydroxybenzoate
Molecular FormulaC9H10O4
InChI KeyLVYLCBNXHHHPSB-UHFFFAOYSA-N
SMILESOCCOC(=O)C1=CC=CC=C1O
さらに表示
Infrared spectrumConforms
GC>=98.0 %
Appearance (Color)Colorless to almost colorless
Appearance (Form)Oily liquid or fused mass
Refractive index1.5480 to 1.5510 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only