Nitronium tetrafluoroborate, 97%
Nitronium tetrafluoroborate, 97%
Nitronium tetrafluoroborate, 97%
Nitronium tetrafluoroborate, 97%
Nitronium tetrafluoroborate, 97%
Thermo Scientific Chemicals

Nitronium tetrafluoroborate, 97%

CAS: 13826-86-3 | BF4NO2 | 132.81 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
Catalog number 174380050
Price (USD)
66.65
Online exclusive
74.30
Save 7.65
Each
Quantity:
5 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
66.65
Online exclusive
74.30
Save 7.65
Each
Chemical Identifiers
CAS13826-86-3
IUPAC Namedioxoamine; tetrafluoroboranuide
Molecular FormulaBF4NO2
InChI KeyXYRQURYIJKJVSF-UHFFFAOYSA-N
SMILES[O-][NH+]=O.F[B-](F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Colorless to white
Assay>=96.0 %
Appearance (Form)Adhering crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only