Tetrafluoroborato de nitronio, 97 %, Thermo Scientific Chemicals
Tetrafluoroborato de nitronio, 97 %, Thermo Scientific Chemicals
Tetrafluoroborato de nitronio, 97 %, Thermo Scientific Chemicals
Tetrafluoroborato de nitronio, 97 %, Thermo Scientific Chemicals
Tetrafluoroborato de nitronio, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tetrafluoroborato de nitronio, 97 %, Thermo Scientific Chemicals

CAS: 13826-86-3 | BF4NO2 | 132.81 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
5 g
25 g
100 g
Envase:
Frasco de plástico
Número de catálogo 174381000
Precio (USD)
-
Cantidad:
100 g
Envase:
Frasco de plástico
Pedido a granel o personalizado
Identificadores químicos
CAS13826-86-3
IUPAC Namedioxoamine; tetrafluoroboranuide
Molecular FormulaBF4NO2
InChI KeyXYRQURYIJKJVSF-UHFFFAOYSA-N
SMILES[O-][NH+]=O.F[B-](F)(F)F
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Colorless to white
Assay>=96.0 %
Appearance (Form)Adhering crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only