PepMap™ Neo Trap Cartridge
PepMap™ Neo Trap Cartridge
PepMap™ Neo Trap Cartridge
Thermo Scientific™

PepMap™ Neo Trap Cartridge

Use the Thermo Scientific™ PepMap™ Neo Trap Cartridge with PepMap™ Neo UHPLC Columns to filter impurities from your peptide mapping samples.
Have Questions?
Catalog NumberLength (Metric)
1745005 mm
Catalog number 174500
Price (USD)
607.27
Pack of 3
Length (Metric):
5 mm
Price (USD)
607.27
Pack of 3

Use the Thermo Scientific™ PepMap™ Neo Trap Cartridge with Double nanoViper™ PepMap™ Neo UHPLC Columns or EASY-Spray™ PepMap™ Neo UHPLC Columns to filter impurities from your peptide mapping samples. Connect the trap cartridge to your HPLC system with the Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, & nanoViper™ Fittings (Catalog Numbers 174501 and 174502).

Specifications
Column FormatTrap Cartridge
Column TypeReversed Phase
DescriptionPepMap™ Neo 5 μm C18 300 μm X 5 mm Trap Cartridge
EndcappedYes
For Use WithPepMap™ Neo Nano Trap Cartridge Holder with PEEK Tubing and nanoViper™ Fittings (part number 174502)
For Use With (Application)Trap and Elute
Max. Pressure22,000 psi (1500 bar)
Packing MaterialSpherical, Fully Porous, Ultrapure Silica
Stationary PhaseC18
Surface Area450 m2/g
Temperature60°C
USP TypeL1
Carbon Load15%
Diameter (Metric)300 μm
Length (Metric)5 mm
Particle Size5 μm
PhaseReversed Phase
Pore Size100 Å
Product LinePepMap Neo
pH2 to 8
Unit SizePack of 3