PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, & nanoViper™ Fittings
Actual product may vary
PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, & nanoViper™ Fittings
PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, & nanoViper™ Fittings
PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, & nanoViper™ Fittings
Thermo Scientific™
PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, & nanoViper™ Fittings
Use the Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, and nanoViper™ Fittings to connect your PepMap™ Neo Trap Cartridge to your HPLC system.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberProduct TypeIncludes
174502Cartridge Holder KitCartridge Holder, Tubing, Fittings
174501TubingTubing, Fittings
Catalog number 174502
Price (USD)
694.57
Each
Product Type:
Cartridge Holder Kit
Includes:
Cartridge Holder, Tubing, Fittings
Price (USD)
694.57
Each
Use the Thermo Scientific™ PepMap™ Neo Trap Cartridge Holder, PEEK Tubing, and nanoViper™ Fittings to connect your PepMap Neo Trap Cartridge (Catalog Number 174500) to your HPLC system.
See Viper and nanoViper resources
Specifications
DescriptionNano Trap Cartridge Holder with PEEK Tubing and nanoViper™ Fittings
For Use WithPepMap™ Neo Trap Cartridge (part number 174500)
For Use With (Application)Trap and Elute, Peptide Mapping
IncludesCartridge Holder, Tubing, Fittings
Max. Pressure22,000 psi (1500 bar)
Product TypeCartridge Holder Kit
QuantityEach
Temperature60°C
Diameter (Metric)30 μm (tubing)
Length (Metric)5 mm
Product LinePepMap Neo
Unit SizeEach