4-Isopropylphenol, 98%
4-Isopropylphenol, 98%
4-Isopropylphenol, 98%
Thermo Scientific Chemicals

4-Isopropylphenol, 98%

CAS: 99-89-8 | C9H12O | 136.19 g/mol
Quantity:
5 g
100 g
250 g
1 kg
Packaging:
Glass bottle
Catalog number 174510010
Price (JPY)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS99-89-8
IUPAC Name4-(propan-2-yl)phenol
Molecular FormulaC9H12O
InChI KeyYQUQWHNMBPIWGK-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)White to pink to brown
Appearance (Form)Powder or crystals or fused solid
Melting point58°C to 63°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only