2,5-Dimethylpyrazine, 99%
2,5-Dimethylpyrazine, 99%
2,5-Dimethylpyrazine, 99%
Thermo Scientific Chemicals

2,5-Dimethylpyrazine, 99%

CAS: 123-32-0 | C6H8N2 | 108.14 g/mol
数量:
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 174520250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS123-32-0
IUPAC Name2,5-dimethylpyrazine
Molecular FormulaC6H8N2
InChI KeyLCZUOKDVTBMCMX-UHFFFAOYSA-N
SMILESCC1=CN=C(C)C=N1
さらに表示
Infrared spectrumConforms
Appearance (Color)Clear colorless to light yellow
Refractive index1.4987 to 1.5007 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only