4-Isopropylaniline, 99%
4-Isopropylaniline, 99%
4-Isopropylaniline, 99%
Thermo Scientific Chemicals

4-Isopropylaniline, 99%

CAS: 99-88-7 | C9H13N | 135.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Packaging:
Glass bottle
Catalog number 174530100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS99-88-7
IUPAC Name4-(propan-2-yl)aniline
Molecular FormulaC9H13N
InChI KeyLRTFPLFDLJYEKT-UHFFFAOYSA-N
SMILESCC(C)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Refractive index1.5420 to 1.5440 (20°C, 589 nm)
Appearance (Form)Liquid
GC>=98.5 %
Appearance (Color)Clear light brown to red-brown
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only