Triethoxyvinylsilane, 97%, Thermo Scientific Chemicals
Triethoxyvinylsilane, 97%, Thermo Scientific Chemicals
Triethoxyvinylsilane, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Triethoxyvinylsilane, 97%, Thermo Scientific Chemicals

CAS: 78-08-0 | Molecular Formula: C8H18O3Si | Molecular Weight: 190.31 g/mol
Have Questions?
Catalog NumberQuantityPackaging
174615000500 gGlass bottle
Catalog number 174615000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS78-08-0
IUPAC Nameethenyltriethoxysilane
Molecular FormulaC8H18O3Si
InChI KeyFWDBOZPQNFPOLF-UHFFFAOYSA-N
SMILESCCO[Si](OCC)(OCC)C=C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Infrared spectrumConforms
Refractive index1.3970 to 1.4020 (20°C, 589 nm)
GC>=96.0 %
Appearance (Form)Liquid

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only