Nunc™ Multidishes with UpCell™ Surface
Thermo Scientific™

Nunc™ Multidishes with UpCell™ Surface

Support non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of WellsCulture Area
174898481.1 sq. cm
174899241.9 sq. cm
174900123.5 sq. cm
17490169.6 sq. cm
Catalog number 174898
Price (USD)
421.65
Online exclusive
449.00
Save 27.35
Case of 6
No. of Wells:
48
Culture Area:
1.1 sq. cm
Price (USD)
421.65
Online exclusive
449.00
Save 27.35
Case of 6

Support non-enzymatic harvesting of adherent cells for preservation of cell viability and surface proteins with the temperature-responsive surface of Thermo Scientific™ Nunc™ UpCell™ Dishes. Simply transfer the culture to room temperature to collect cells in suspension or in the form of cell sheet with the help of a supportive membrane.

Features:
  • The hydrophicility of the UpCell surface changes with the external temperature, allowing cells to detach
  • No need for dissociation enzymes, ensuring intact cell surface proteins
  • No need for physical scraping, resulting in high cell viability
  • Supportive membranes included in the 6-well plates allow for harvesting of the cell sheets
  • Customized bar coding available
  • Shelf Life: 3 years from date of manufacturing

Recommended for:

  • Culture passaging
  • Subsequent single-cell analyses (e.g. flow cytometry)
  • 3D tissue modeling for transplantation
  • For research use only
Warranty: 90 days

 

WARNING: Cancer – www.P65Warnings.ca.gov
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. For research and single-use only
Specifications
ColorClear
Description48 well, UpCell
No. per Case6
No. per Pack1
Surface TreatmentUpCell, Temperature Responsive
Well ShapeRectangular, Flat Bottom (F-well)
Culture Area1.1 sq. cm
No. of Wells48
SterilitySterile
TypeMulti Dish
Volume (Metric) Well0.5 mL (Well)
Volume (Metric) Working0.5 mL
Unit SizeCase of 6

Frequently asked questions (FAQs)

Why does temperature reduction detach cells from the Nunc dishes with UpCell Surface?

The UpCell Surface is composed of a covalently bound polymer that changes from being slightly hydrophobic to being very hydrophilic when the temperature is reduced to less than 32 degrees C. This results in the binding of water to the surface, swelling of the surface, and finally the release of the cells.

In cultureware with UpCell surface, does the temperature-responsive polymer detach with the cells?

The temperature-responsive polymer is covalently bound to the polystyrene dish or plate and will not detach from the product under normal cell culture procedures and conditions. The products have successfully passed the USP biological reactivity class VI testing.

What is special about the "UpCell surface with grid"?

The “UpCell surface with grid” products have a 3 x 3 mm grid design over the entire culture surface. Cell attachment and growth is restricted to within these small squares, preventing cells from clumping, and enabling easier harvest of the cells as single cells or small cell clusters. Products with “UpCell surface with grid” are suitable for the preparation of cells for flow cytometry analysis but cannot be used for cell-sheet applications. We offer 60 mm and 100 mm dishes with “UpCell surface with grid” whereas the 35 mm dishes contain supportive membrane for harvesting of cell sheets.

Which cell types can I use with the UpCell surface?

You can use the UpCell Surface with many cell types, including fibroblasts, mesenchymal stem cells, myocytes, keratinocytes, macrophages, and different epithelium and tumor cells. If cells do not detach spontaneously from the UpCell surface by lowering of the temperature to below 32 degrees C, the cells can be detached by flushing the surface with medium or by tapping on the side of the dish or plate. Cells that produce a lot of extracellular matrix and attach very strongly to the UpCell surface may be difficult to detach by temperature reduction alone. For these cell types, the simultaneous use of diluted trypsin is recommended.

Can I coat the UpCell surface with biomolecules to enable cell attachment?

Coating of the UpCell surface with biomolecules, for example, collagen, fibronectin, poly-lysine and laminin, is possible and should be done if you experience difficulties in attaching your cells to the UpCell surface. The coating procedure should be performed at 37 degrees C.