Iodomethane-d3, for NMR, 99+ atom % D
Iodomethane-d3, for NMR, 99+ atom % D
Iodomethane-d3, for NMR, 99+ atom % D
Thermo Scientific Chemicals

Iodomethane-d3, for NMR, 99+ atom % D

CAS: 865-50-9 | CH3I | 144.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 174950050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS865-50-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
D-Enrichment>=99.0 %
Water=<0.05 % (Coulometric)
Appearance (Color)Clear colorless to slightly yellowish to
Appearance (Color)reddish
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
RUO – Research Use Only