Borane-tetrahydrofuran complex, 1M solution in THF, Stabilized, AcroSeal™
Borane-tetrahydrofuran complex, 1M solution in THF, Stabilized, AcroSeal™
Borane-tetrahydrofuran complex, 1M solution in THF, Stabilized, AcroSeal™
Borane-tetrahydrofuran complex, 1M solution in THF, Stabilized, AcroSeal™
Thermo Scientific Chemicals

Borane-tetrahydrofuran complex, 1M solution in THF, Stabilized, AcroSeal™

CAS: 14044-65-6 | C4H11BO | 85.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Catalog number 175081000
Price (USD)
79.10
Each
Quantity:
100 mL
Request bulk or custom format
Price (USD)
79.10
Each
Chemical Identifiers
CAS14044-65-6
IUPAC Nameoxolane borane
Molecular FormulaC4H11BO
InChI KeyRMCYTHFAWCWRFA-UHFFFAOYSA-N
SMILESB.C1CCOC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearanceinert atmosphere
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagent
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only