Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Thermo Scientific™ Nalgene™ syringe filters are built to meet your lab’s clarification and sterilization needs. Our filters are available in a variety of sizes and membranes that can be easily used for both sterile and non-sterile laboratory applications. Whether your research necessitates prefiltration, sample preparation, sterile filtration, laboratory filtration, or gas venting, Nalgene syringe filters are lab essentials.
Color Code | Green |
Description | Green; Membrane: nylon; 0.45um pore |
Material (Membrane) | Nylon |
Pore Size | 0.45 μm |
Sterility | Non-sterile |
For Use With (Application) | Sterilizing cryoprotectants such as DMSO and with |
Housing Material | Polypropylene |
Product Line | Nalgene™ |
Type | Syringe Filter |
Volume (Metric) | 10 to 100 mL (Sample) |
Diameter (Metric) | 4 mm |
Diameter (Metric) Membrane | 4 mm |
Unit Size | Pack of 100 |
Material | Polypropylene |
Material (Membrane) | PTFE |
Pore Size | 0.2 μm |
Sterility | Non-sterile |
For Use With (Application) | Laboratory Application |
Housing Material | Polypropylene |
No. per Case | 300 |
Product Line | Nalgene™ |
Type | Syringe Filter |
Volume (Metric) | 10 to 100 mL (Sample) |
Diameter (Metric) | 25 mm |
Unit Size | Case of 300 |
Filter Type | Membrane Filter |
Material (Membrane) | PTFE |
Pore Size | 0.2 μm |
Sterility | Non-sterile |
For Use With (Application) | Sterile venting of air or other gasses, sample preparation of samples for instrument analysis such as HPLC and CG |
Housing Material | Polypropylene (Housing) |
Quantity | 100 |
Type | Syringe Filter |
Volume (Metric) | 2 to 10 mL |
Diameter (Metric) | 13 mm |
Unit Size | Case of 100 |
Filter Type | Membrane Filter |
Material (Membrane) | PTFE |
Pore Size | 0.45 μm |
Sterility | Non-sterile |
For Use With (Application) | Venting of air or other gasses, sample preparation of samples for instrument analysis such as HPLC and CG |
Housing Material | Polypropylene (Housing) |
Quantity | 100 |
Type | Syringe Filter |
Volume (Metric) | 2 to 10 mL |
Diameter (Metric) | 13 mm |
Unit Size | Case of 100 |
Material (Membrane) | Glass Fiber |
Pore Size | 1.1 μm |
Sterility | Non-sterile |
For Use With (Application) | Laboratory Application |
Housing Material | Polypropylene |
No. per Case | 50 |
No. per Pack | 50 |
Product Line | Nalgene™ |
Type | Syringe Filter |
Volume (Metric) | 10 to 100 mL (Sample) |
Diameter (Metric) | 25 mm |
Diameter (Metric) Membrane | 25 mm |
Unit Size | Case of 50 |
Material (Membrane) | SFCA |
Pore Size | 0.2 μm |
Sterility | Non-sterile |
For Use With (Application) | Filtration of biological/immunological samples, cell culture fluids, cellular solutions, and biological fluids |
Green Features | Fewer resources used and less waste |
Housing Material | Polypropylene (Housing) |
Product Line | Nalgene™ |
Type | Syringe Filter |
Volume (Metric) | 10 to 100 mL (Sample) |
Diameter (Metric) | 25 mm |
Diameter (Metric) Membrane | 25 mm |
Unit Size | Case of 500 |
Material (Membrane) | SFCA |
Pore Size | 0.2 μm |
Sterility | Non-sterile |
For Use With (Application) | Filtration of biological/immunological samples, cell culture fluids, cellular solutions, and biological fluids |
Green Features | Fewer resources used and less waste |
Housing Material | Polypropylene (Housing) |
Product Line | Nalgene™ |
Type | Syringe Filter |
Volume (Metric) | 10 to 100 mL (Sample) |
Diameter (Metric) | 25 mm |
Diameter (Metric) Membrane | 25 mm |
Unit Size | Pack of 100 |
Material (Membrane) | SFCA |
Pore Size | 0.45 μm |
Sterility | Non-sterile |
For Use With (Application) | Filtration of biological/immunological samples, cell culture fluids, cellular solutions, and biological fluids |
Green Features | Fewer resources used and less waste |
Housing Material | Polypropylene (Housing) |
Product Line | Nalgene™ |
Type | Syringe Filter |
Volume (Metric) | 10 to 100 mL (Sample) |
Diameter (Metric) | 25 mm |
Diameter (Metric) Membrane | 25 mm |
Unit Size | Case of 500 |
Material (Membrane) | SFCA |
Pore Size | 0.45 μm |
Sterility | Non-sterile |
For Use With (Application) | Filtration of biological/immunological samples, cell culture fluids, cellular solutions, and biological fluids |
Green Features | Fewer resources used and less waste |
Housing Material | Polypropylene (Housing) |
Product Line | Nalgene™ |
Type | Syringe Filter |
Volume (Metric) | 10 to 100 mL (Sample) |
Diameter (Metric) | 25 mm |
Diameter (Metric) Membrane | 25 mm |
Unit Size | Pack of 100 |
Material (Membrane) | Cellulose Acetate |
Pore Size | 0.8 μm |
Sterility | Non-sterile |
For Use With (Application) | Filtration of buffers, aqueous solutions and reagents |
Green Features | Fewer resources used and less waste |
Housing Material | Polypropylene (Housing) |
Product Line | Nalgene™ |
Type | Syringe Filter |
Volume (Metric) | 10 to 100 mL (Sample) |
Diameter (Metric) | 25 mm |
Diameter (Metric) Membrane | 25 mm |
Unit Size | Case of 500 |
Catalog Number | Specifications | Unit Size | Diameter (Metric) | Material (Membrane) | Pore Size | Sterility | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
176-0045PK also known as 176-0045 | Pack of 100 | 4 mm | Nylon | 0.45 μm | Non-sterile | Price: 309.30 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
199-2020 | Case of 300 | 25 mm | PTFE | 0.2 μm | Non-sterile | Price: 1,476.00 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
721-1320 | Case of 100 | 13 mm | PTFE | 0.2 μm | Non-sterile | Price: 346.00 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
721-1345 | Case of 100 | 13 mm | PTFE | 0.45 μm | Non-sterile | Price: 346.00 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
722-2000 | Case of 50 | 25 mm | Glass Fiber | 1.1 μm | Non-sterile | Price: 204.00 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
724-2020 | Case of 500 | 25 mm | SFCA | 0.2 μm | Non-sterile | Price: 2,012.00 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
724-2020PK also known as 724-2020 | Pack of 100 | 25 mm | SFCA | 0.2 μm | Non-sterile | Price: 434.50 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
724-2045 | Case of 500 | 25 mm | SFCA | 0.45 μm | Non-sterile | Price: 2,012.00 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
724-2045PK also known as 724-2045 | Pack of 100 | 25 mm | SFCA | 0.45 μm | Non-sterile | Price: 434.50 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
724-2080 | Case of 500 | 25 mm | Cellulose Acetate | 0.8 μm | Non-sterile | Price: 2,119.00 Your price: | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Surfactant-free Cellulose Acetate (SFCA)
Cellulose Acetate (CA)
Nylon