Thermo Scientific™
Specialized Clamps and Accessories
Related applications:
Anatomical Pathology
Ensure accuracy and efficiency with Thermo Scientific™ Specialized Clamps and Accessories.
Back to top
  • For clamping thin materials
  • Spring-loaded jaws secure specimen
  • Compensation slot ensures correct specimen clamping
  • Orienting on the Z-axis
  • Foil clamp is fitted directly on the specimen cylinder
  • Sandwich supporting material is for foil clamp to section thin foil specimens; 50 squares of hard material and two strips