2,5-Dimethylfuran, 99%
2,5-Dimethylfuran, 99%
2,5-Dimethylfuran, 99%
Thermo Scientific Chemicals

2,5-Dimethylfuran, 99%

CAS: 625-86-5 | C6H8O | 96.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
100 mL
500 mL
Packaging:
View all
Glass Bottle
Catalog number 176035000
Price (EUR)
463,00
Each
Quantity:
500 mL
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
463,00
Each
Chemical Identifiers
CAS625-86-5
IUPAC Name2,5-dimethylfuran
Molecular FormulaC6H8O
InChI KeyGSNUFIFRDBKVIE-UHFFFAOYSA-N
SMILESCC1=CC=C(C)O1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Refractive index1.4400 to 1.4440 (20°C, 589 nm)
Appearance (Color)Clear colorless to amber
AppearanceMay darken on storage
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only