4-(2-Pyridylazo)resorcinol monosodium salt monohydrate, 99+%
4-(2-Pyridylazo)resorcinol monosodium salt monohydrate, 99+%
4-(2-Pyridylazo)resorcinol monosodium salt monohydrate, 99+%
Acros Organics

4-(2-Pyridylazo)resorcinol monosodium salt monohydrate, 99+%

CAS: 16593-81-0 | C11H9N3O2 | 215.21 g/mol
Have Questions?
Catalog NumberQuantityPackaging
176321000
also known as 17632
100 gGlass bottle
Catalog number 176321000
also known as 17632
HSN 29333919
Price (INR)
129,207.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
129,207.00
Each
Chemical Identifiers
CAS16593-81-0
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with HClO4>=99 % (on dry substance)
Water=<10 % (K.F.)
Suitability(for Pb determination) Passes test
Solubility(0.1 % in water) Clear
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only