Boron trichloride, 1M solution in methylene chloride, AcroSeal™
Boron trichloride, 1M solution in methylene chloride, AcroSeal™
Boron trichloride, 1M solution in methylene chloride, AcroSeal™
Thermo Scientific Chemicals

Boron trichloride, 1M solution in methylene chloride, AcroSeal™

CAS: 10294-34-5 | BCl3 | 117.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
800 mL
Packaging:
View all
AcroSeal™ Glass bottle
Catalog number 176681000
Price (USD)
111.00
Each
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Request bulk or custom format
Price (USD)
111.00
Each
Chemical Identifiers
CAS10294-34-5
IUPAC Nametrichloroborane
Molecular FormulaBCl3
InChI KeyFAQYAMRNWDIXMY-UHFFFAOYSA-N
SMILESClB(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to brown
Appearance (Form)Solution
AppearanceMay darken on storage
Titration Argentometric0.9 to 1.1 M
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only