Cyclohexane, 99.5%, for analysis
Cyclohexane, 99.5%, for analysis
Cyclohexane, 99.5%, for analysis
Thermo Scientific Chemicals

Cyclohexane, 99.5%, for analysis

CAS: 110-82-7 | C6H12 | 84.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
5 L
25 L
Packaging:
View all
Glass Bottle
Catalog number 176810025
Price (USD)
136.65
Online exclusive
152.00
Save 15.35
Each
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
136.65
Online exclusive
152.00
Save 15.35
Each
Chemical Identifiers
CAS110-82-7
IUPAC Namecyclohexane
Molecular FormulaC6H12
InChI KeyXDTMQSROBMDMFD-UHFFFAOYSA-N
SMILESC1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Color scale=<10 APHA
Water=<0.05 %
Zinc (Zn)=<0.1 ppm
Sodium (Na)=<1 ppm
Residue after evaporation=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
RUO – Research Use Only