Triethylborane, 1M solution in THF, AcroSeal™
Triethylborane, 1M solution in THF, AcroSeal™
Triethylborane, 1M solution in THF, AcroSeal™
Thermo Scientific Chemicals

Triethylborane, 1M solution in THF, AcroSeal™

CAS: 97-94-9 | C6H15B | 98.00 g/mol
数量:
100 mL
800 mL
パッケージ:
すべて表示
AcroSeal™ Glass bottle
製品番号(カタログ番号) 176981000
価格(JPY)
-
数量:
100 mL
パッケージ:
AcroSeal™ Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS97-94-9
IUPAC Nametriethylborane
Molecular FormulaC6H15B
InChI KeyLALRXNPLTWZJIJ-UHFFFAOYSA-N
SMILESCCB(CC)CC
さらに表示
Appearance (Color)Colorless to amber
Strength0.95 to 1.30 M
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Lewis acid
RUO – Research Use Only