Borane-pyridine complex, 95%
Borane-pyridine complex, 95%
Borane-pyridine complex, 95%
Thermo Scientific Chemicals

Borane-pyridine complex, 95%

CAS: 110-51-0 | C5H8BN | 92.94 g/mol
Have Questions?
Catalog NumberQuantity
17701025025 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 177010250
Price (USD)
87.20
Each
Quantity:
25 mL
Request bulk or custom format
Price (USD)
87.20
Each
Chemical Identifiers
CAS110-51-0
IUPAC Name(pyridin-1-ium-1-yl)boranuide
Molecular FormulaC5H8BN
InChI KeyLPGWNCNRGQANGC-UHFFFAOYSA-N
SMILES[BH3-][N+]1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear to slightly hazy colorless to amber
AppearanceMay darken on storage
Infrared spectrumConforms
Titration>=94 % (Active hydrogen)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only