Hepatocyte Wash Medium
Hepatocyte Wash Medium
Hepatocyte Wash Medium
Gibco™

Hepatocyte Wash Medium

Hepatocyte Wash Medium
Have Questions?
카탈로그 번호수량
17704024500 mL
카탈로그 번호 17704024
제품 가격(KRW)
151,000
온라인 행사
Ends: 31-Dec-2026
167,000
할인액 16,000 (10%)
Each
수량:
500 mL
Customize this product
제품 가격(KRW)
151,000
온라인 행사
Ends: 31-Dec-2026
167,000
할인액 16,000 (10%)
Each

Hepatocyte Wash Medium is an enriched Williams' Medium E for the reduction of cell membrane leakage, ease of centrifugation and high viability.

Hepatocyte Wash Medium는 세포막 누출은 낮추고 원심분리를 쉽게 하며 생존율을 높이기 위해 사용하는 농축된 Williams' Medium E입니다.

For Research Use Only. Not for use in diagnostic procedures.

사양
제품 유형Hepatocyte Wash Medium
수량500 mL
배송 조건Room Temperature
형태Liquid
Unit SizeEach
구성 및 보관
Store in refrigerator (2–8°C) and protect from light.

자주 묻는 질문(FAQ)

How long can I keep my media after supplementing with serum?

Generally speaking, media can be used for up to three weeks after supplementation with serum. There are no formal studies to support this, but it is the rule of thumb used by our scientists.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

My medium was shipped at room temperature but it is supposed to be stored refrigerated. Is it okay?

We routinely ship media that require long-term storage in the refrigerator at room temperature. We have done studies on representative media formulations to show that media can be at room temperature for up to a week without a problem.

Find additional tips, troubleshooting help, and resources within our Mammalian Cell Culture Basics Support Center.

How can I remove mycoplasma contamination from my cell culture medium?

Very often mycoplasma contamination cannot be removed from the culture so it should be discarded. You may have a unique culture that you prefer not to discard and would like to try to clean it. Ciprofloxacin and Plasmocin have reportedly been used for this application. If interested in a protocol or directions for use, check with the antibiotic supplier or published literature. Note that mycoplasma are very difficult to remove from culture and spread easily so the treated cultures should be quarantined until clear of mycoplasma, and your laboratory should be thoroughly cleaned.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

I see a decrease in growth of my culture. What should I do?

Try changing the medium or serum. Compare media formulations for differences in glucose, amino acids, and other components. Compare an old lot of serum with a new lot. Increase initial cell inoculums. Lastly, adapt cells sequentially to new medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

My cells are not adhering to the culture vessel. What should I do?

This can occur if cells are overly trypsinized. Trypsinize for a shorter time or use less trypsin. Mycoplasma contamination could also cause this problem. Segregate your culture and test for mycoplasma infection. Lastly, check for attachment factors in the medium.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.