Cottonseed oil, pure
Thermo Scientific Chemicals

Cottonseed oil, pure

CAS: 8001-29-4
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Catalog number 177115000
Price (EUR)
44,65
Online Exclusive
49,10
Save 4,45 (9%)
Each
Quantity:
500 mL
Request bulk or custom format
Price (EUR)
44,65
Online Exclusive
49,10
Save 4,45 (9%)
Each
Chemical Identifiers
CAS8001-29-4
CAS8001-29-4
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4720 to 1.4730 (20°C, 589 nm)
Composition of fatty acids25 % (Palmitic acid) (typical)
Composition of fatty acids16 to 24 % (Oleic acid) (typical)
Composition of fatty acids50 to 55 % (Linoleic acid) (typical)
Composition of fatty acids=<1.5 % (Linolenic acid and higher) (typical)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only