D(+)-Sucrose, 99.7%, for biochemistry
D(+)-Sucrose, 99.7%, for biochemistry
D(+)-Sucrose, 99.7%, for biochemistry
Thermo Scientific Chemicals

D(+)-Sucrose, 99.7%, for biochemistry

D(+)-Sucrose, CAS # 57-50-1, is a disaccharide used in a variety of biochemical applications such as protein purification. It is also known as sugar, sucrose, alpha-D-glucopyranoside, and D-(+)-saccharose. | CAS: 57-50-1 | C12H22O11 | 342.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 177140010
Price (USD)
-
Quantity:
1 kg
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS57-50-1
IUPAC Name(2R,3R,4S,5S,6R)-2-{[(2S,3S,4S,5R)-3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC12H22O11
InChI KeyCZMRCDWAGMRECN-PWPRYFECNA-N
SMILESOC[C@H]1O[C@@](CO)(O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Iron (Fe)=<0.5 ppm
Sulfite (as SO2)=<15 ppm
Reducing sugars=<0.04 %
Insoluble matter=<0.0007 % (in water)
pH6.5 to 7.5 (10 % solution)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • D(+)-Sucrose is a disaccharide carbohydrate
  • It is also known as sugar, sucrose, alpha-D-glucopyranoside, and D-(+)-saccharose

Applications

  • D(+)-Sucrose can be used for protein and nucleic acid purification
  • Its fatty acid esters have laboratory research applications in transdermal systems
  • It is also used for sucrose-gradient centrifugation, which may be employed for the selective isolation of Nocardia species from soil
RUO – Research Use Only