Toluene, 99+%, extra pure
Toluene, 99+%, extra pure
Toluene, 99+%, extra pure
Thermo Scientific Chemicals

Toluene, 99+%, extra pure

CAS: 108-88-3 | C7H8 | 92.14 g/mol
Quantity:
2.5 L
5 L
10 L
25 L
Packaging:
View all
Metal Drum
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 177160250
Price (JPY)
-
Quantity:
25 L
Packaging:
Metal Drum
Request bulk or custom format
Chemical Identifiers
CAS108-88-3
IUPAC Nametoluene
Molecular FormulaC7H8
InChI KeyYXFVVABEGXRONW-UHFFFAOYSA-N
SMILESCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after evaporation=<0.003 %
GC>=99.0 %
Water=<0.05 % (K.F.)
Appearance (Form)Clear liquid
Color scale=<10 APHA
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only