Carbon disulfide, 99.9%, extra pure, low in benzene content
Carbon disulfide, 99.9%, extra pure, low in benzene content
Carbon disulfide, 99.9%, extra pure, low in benzene content
Thermo Scientific Chemicals

Carbon disulfide, 99.9%, extra pure, low in benzene content

CAS: 75-15-0 | CS2 | 76.131 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number 177280010
Price (EUR)
115,65
Online exclusive
128,00
Save 12,35
Each
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (EUR)
115,65
Online exclusive
128,00
Save 12,35
Each
Chemical Identifiers
CAS75-15-0
IUPAC Namemethanedithione
Molecular FormulaCS2
InChI KeyQGJOPFRUJISHPQ-UHFFFAOYSA-N
SMILESS=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Assay>=99.85 % (By difference)
Residue after evaporation=<20 ppm
Hydrogen sulfide (H2S)=<3 ppm
Appearance (Color)Clear colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only