Difenil diselenida, 99 %, Thermo Scientific Chemicals
Difenil diselenida, 99 %, Thermo Scientific Chemicals
Difenil diselenida, 99 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Difenil diselenida, 99 %, Thermo Scientific Chemicals

CAS: 1666-13-3 | C12H10Se2 | 312.15 g/mol
Have Questions?
Número de catálogoCantidadEnvase
17757025025 gFrasco de vidrio
Número de catálogo 177570250
Precio (USD)
-
Cantidad:
25 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS1666-13-3
IUPAC Name(phenyldiselanyl)benzene
Molecular FormulaC12H10Se2
InChI KeyYWWZCHLUQSHMCL-UHFFFAOYSA-N
SMILES[Se]([Se]C1=CC=CC=C1)C1=CC=CC=C1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Infrared spectrumConforms
GC>=98.0 %
Appearance (Color)Yellow
Appearance (Form)Crystalline powder
Melting point59.0°C to 64.0°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only