Cellulose triacetate
Cellulose triacetate
Cellulose triacetate
Thermo Scientific Chemicals

Cellulose triacetate

CAS: 9012-09-3 | (C24H32O16)n
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
500 g
Packaging:
Plastic bottle
Catalog number 177825000
Price (USD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS9012-09-3
Molecular Formula(C24H32O16)n
SMILESCC(=O)OC[C@H]1O[C@@H](O[C@@H]2[C@@H](COC(C)=O)OC(-*)[C@H](OC(C)=O)[C@H]2OC(C)=O)[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@@H]1O-*
Molecular Weight (g/mol)NaN
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Acetyl content43 to 44 wt %
Infrared spectrumConforms
Appearance (Form)Pellets or rods
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only