Ácido poliacrílico, patrón secundario, P.M. medio 1 080 000 N.M. medio 135 000, Thermo Scientific Chemicals
Ácido poliacrílico, patrón secundario, P.M. medio 1 080 000 N.M. medio 135 000, Thermo Scientific Chemicals
Ácido poliacrílico, patrón secundario, P.M. medio 1 080 000 N.M. medio 135 000, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ácido poliacrílico, patrón secundario, P.M. medio 1 080 000 N.M. medio 135 000, Thermo Scientific Chemicals

CAS: 9003-01-4 | (C3H4O2)n | 72.06 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
1 g
10 g
Envase:
Frasco de vidrio
Número de catálogo 178060010
Precio (EUR)
44,65
Precio exclusivo compra web
49,00
Ahorro 4,35 (9%)
Each
Cantidad:
1 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Precio (EUR)
44,65
Precio exclusivo compra web
49,00
Ahorro 4,35 (9%)
Each
Identificadores químicos
CAS9003-01-4
IUPAC NamePoly(acrylic acid)
Molecular Formula(C3H4O2)n
SMILESOC(=O)C(-*)C-*
MDL NumberMFCD00084394
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Form)Powder
Infrared spectrumConforms
Loss on drying=<10 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only