Agarose I (Molecular Biology Grade)
Thermo Scientific™

Agarose I (Molecular Biology Grade)

Thermo Scientific Pierce Agaroses are highly purified and carefully blended formulations of regular- or low-melting agarose that provide uniform lot-to-lotRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
17852500 g
17850100 g
Catalog number 17852
Price (USD)
1,196.00
Each
Quantity:
500 g
Price (USD)
1,196.00
Each
Thermo Scientific Pierce Agaroses are highly purified and carefully blended formulations of regular- or low-melting agarose that provide uniform lot-to-lot consistency for preparation of electrophoresis gels.

Features:

• A regular melting temperature agarose for electrophoresis requiring an exceptional level of purity with unequaled performance

Related Products
Agarose II (Low Melt)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Gel Electrophoresis
Melting PointStandard Melting Point
Product TypeAgarose I (Molecular Biology Grade)
Quantity500 g
Separation Range100 bp to >30 kb
Unit SizeEach
Contents & Storage
Store at room temperature.