Poly(styrene), average M.W. 260,000
Poly(styrene), average M.W. 260,000
Poly(styrene), average M.W. 260,000
Thermo Scientific Chemicals

Poly(styrene), average M.W. 260,000

CAS: 9003-53-6 | (C8H8)n | 104.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
1 kg
Packaging:
Glass bottle
Catalog number 178890250
Price (USD)
21.65
Online exclusive
24.00
Save 2.35
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
21.65
Online exclusive
24.00
Save 2.35
Each
Chemical Identifiers
CAS9003-53-6
Molecular Formula(C8H8)n
SMILES*-CC(-*)C1=CC=CC=C1
MDL NumberMFCD00084450
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Pellets
Appearance (Color)Clear
Infrared spectrumConforms
Loss on drying=<1 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only