Polysulfone Resin, Pellets
Polysulfone Resin, Pellets
Polysulfone Resin, Pellets
Thermo Scientific Chemicals

Polysulfone Resin, Pellets

CAS: 25135-51-7 | (C27H22O4S)n | NaN g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
250 g
Packaging:
View all
Plastic bottle
Catalog number 178912500
Price (USD)
189.65
Online Exclusive
210.00
Save 20.35 (10%)
Each
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
189.65
Online Exclusive
210.00
Save 20.35 (10%)
Each
Chemical Identifiers
CAS25135-51-7
Molecular Formula(C27H22O4S)n
SMILESCC(C)(C1=CC=C(O-*)C=C1)C1=CC=C(OC2=CC=C(C=C2)S(=O)(=O)C2=CC=C(-*)C=C2)C=C1
MDL NumberMFCD00134387
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Amber to beige
Appearance (Form)Pellets
Loss on drying=<1 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only