Ethidium Bromide Solution (10 mg/mL)
Thermo Scientific™

Ethidium Bromide Solution (10 mg/mL)

Thermo Scientific Pierce Ethidium Bromide is available as a concentrated (10 mg/mL) solution to accommodate most molecular biology applications forRead more
Have Questions?
Catalog NumberQuantity
178985 x 10 mL
Catalog number 17898
Price (USD)
339.00
Each
Quantity:
5 x 10 mL
Request bulk or custom format
Price (USD)
339.00
Each
Thermo Scientific Pierce Ethidium Bromide is available as a concentrated (10 mg/mL) solution to accommodate most molecular biology applications for fluorescent DNA visualization.

Features of Ethidium Bromide Solution:

• A safe and convenient way to use ethidium bromide, while minimizing exposure

Related Products
Ethidium Bromide Solution (0.625 mg/mL)
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Detection LocationIn-Gel Detection
Detection MethodFluorescence
Quantity5 x 10 mL
Target MoleculeRNA, DNA
Label or DyeEthidium Bromide
Product TypeEthidium Bromide Solution
Unit SizeEach
Contents & Storage
Store at room temperature, protected from light.

Frequently asked questions (FAQs)

Do you have a protocol for ethidium bromide staining in agarose gels?

For use in agarose gels:

- Add ethidium bromide to melted agarose to a final concentration of 0.5 µg/mL. Do not melt agarose that already contains ethidium bromide.

For staining agarose gels after electrophoresis:

- You can stain gels that have been run in the absence of ethidium bromide by covering the gel in 0.5 µg/mL ethidium bromide in water and gently agitating for 10 to 30 minutes.
- If necessary, gels can be destained by shaking in H2O for an additional 30 minutes. To stain RNA gels, you should minimize staining time and definitely include a destaining period.

What can ethidium bromide be used to detect and what is its sensitivity?

Ethidium bromide can be used to detect ssDNA, RNA, and dsDNA. This fluorescent dye intercalates between the stacked bases of nucleic acids, and exhibits an increased fluorescence at 590 nm. Ethidium bromide can detect down to ~1-10 ng/band of dsDNA in an agarose gel.