Thermo Scientific Pierce Proteinase K is a protease that cleaves at the carboxyl side of aliphatic, aromatic or hydrophobic residues자세히 알아보기
Have Questions?
카탈로그 번호
수량
17916
100 mg
카탈로그 번호 17916
제품 가격(KRW)
248,000
온라인 행사
Ends: 30-Jun-2026
283,000
할인액 35,000 (12%)
Each
수량:
100 mg
대량 주문 또는 맞춤형 요청
제품 가격(KRW)
248,000
온라인 행사
Ends: 30-Jun-2026
283,000
할인액 35,000 (12%)
Each
Thermo Scientific Pierce Proteinase K is a protease that cleaves at the carboxyl side of aliphatic, aromatic or hydrophobic residues and is commonly used to digest and inactivate DNase and RNase in nucleic acid purification.
Proteinase K is a subtilisin-like endolytic protease that is isolated from the saprophytic fungus Tritirachium album. It has a high activity that is stable across a wide range of pH and temperature conditions and is suited to short digestion times. The activity of proteinase K is increased at elevated temperatures up to 65°C. Calcium is not essential to the function of proteinase K. Therefore, EDTA and other chelating agents do not interfere with the activity and may be used alongside proteinase K to inactivate calcium-dependent nucleases in DNA and RNA preparation.
For Research Use Only. Not for use in diagnostic procedures.
사양
형태Powder
수량100 mg
제품 유형Proteinase K
Unit SizeEach
구성 및 보관
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -20°C.