Zein, purified
Zein, purified
Zein, purified
Thermo Scientific Chemicals

Zein, purified

CAS: 9010-66-6
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 179311000
Price (EUR)
53,65
Online exclusive
59,40
Save 5,75 (10%)
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
53,65
Online exclusive
59,40
Save 5,75 (10%)
Each
Chemical Identifiers
CAS9010-66-6
MDL NumberMFCD00062414
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow to straw yellow to tan
Infrared spectrumConforms
Loss on drying=<8.0 % (105°C, 2 h)
Appearance (Form)Powder
Residue after ignition=<2.0 % (800°C)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only