Dibutyltin oxide, 98%
Dibutyltin oxide, 98%
Dibutyltin oxide, 98%
Thermo Scientific Chemicals

Dibutyltin oxide, 98%

CAS: 818-08-6 | C8H18OSn | 248.92 g/mol
Have Questions?
變更視圖buttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
產品號碼 179361000
價格 (TWD)
-
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
化學識別
CAS818-08-6
IUPAC Namedibutylstannanone
Molecular FormulaC8H18OSn
InChI KeyJGFBRKRYDCGYKD-UHFFFAOYSA-N
SMILESCCCC[Sn](=O)CCCC
檢視更多
規格Specification Sheet規格表
Infrared spectrumConforms
Volatile matter=<0.5 %
Chloride (Cl)=<0.2 %
Appearance (Color)White
Appearance (Form)Powder or granular powder
檢視更多
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only