Dibutyltin oxide, 98%
Dibutyltin oxide, 98%
Dibutyltin oxide, 98%
Thermo Scientific Chemicals

Dibutyltin oxide, 98%

CAS: 818-08-6 | C8H18OSn | 248.92 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 179365000
Price (EUR)
103,00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
103,00
Each
Chemical Identifiers
CAS818-08-6
IUPAC Namedibutylstannanone
Molecular FormulaC8H18OSn
InChI KeyJGFBRKRYDCGYKD-UHFFFAOYSA-N
SMILESCCCC[Sn](=O)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or granular powder
Tin (Sn)>=47 %
Infrared spectrumConforms
Volatile matter=<0.5 %
Chloride (Cl)=<0.2 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only