(+)-Limonene, 96%, unstabilized
(+)-Limonene, 96%, unstabilized
(+)-Limonene, 96%, unstabilized
Thermo Scientific Chemicals

(+)-Limonene, 96%, unstabilized

CAS: 5989-27-5 | C10H16 | 136.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
500 mL
2.5 L
Packaging:
View all
Glass bottle
Catalog number 179390050
Price (USD)
-
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5989-27-5
IUPAC Name(4R)-1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene
Molecular FormulaC10H16
InChI KeyXMGQYMWWDOXHJM-JTQLQIEISA-N
SMILESCC(=C)[C@@H]1CCC(C)=CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Specific gravity(20°C) 0.835 to 0.850
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only