Cloruro de 3,3-dimetilacriloilo, 97 %, Thermo Scientific Chemicals
Cloruro de 3,3-dimetilacriloilo, 97 %, Thermo Scientific Chemicals
Cloruro de 3,3-dimetilacriloilo, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cloruro de 3,3-dimetilacriloilo, 97 %, Thermo Scientific Chemicals

CAS: 3350-78-5 | C5H7ClO | 118.56 g/mol
Have Questions?
Número de catálogoCantidadEnvase
1797300505 gFrasco de vidrio
Número de catálogo 179730050
Precio (USD)
-
Cantidad:
5 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS3350-78-5
IUPAC Name3-methylbut-2-enoyl chloride
Molecular FormulaC5H7ClO
InChI KeyBDUBTLFQHNYXPC-UHFFFAOYSA-N
SMILESCC(C)=CC(Cl)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
GC>=96.0 %
Appearance (Color)Clear colorless to yellow
AppearanceMay darken on storage
Infrared spectrumConforms
Refractive index1.4750 to 1.4780 (20°C, 589 nm)
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only