Product Image
Invitrogen™

eBioscience™ Avidin HRP

The avidin horseradish peroxidase is commonly used as a second step for detection of biotinylated antibodies by ELISA and westernRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
18-4100-94500 μL
18-4100-511 mL
Catalog number 18-4100-94
Price (MXN)
-
Quantity:
500 μL
The avidin horseradish peroxidase is commonly used as a second step for detection of biotinylated antibodies by ELISA and western blot. Avidin binds to biotin with high affinity. [Note: Sodium azide is an inhibitor of horseradish peroxidase enzymatic activity and should not be added to the AV-HRP diluent.]

Conjugate
HRP

Reported Application
Western Blotting, ELISA

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Label or DyeHRP (Horseradish Peroxidase)
Quantity500 μL
Target MoleculeBiotin
FormLiquid
Product LineeBioscience
Unit SizeEach