CD30 (Ki-1) Mouse mAb (clone Ber-H2), concentrate

Catalog number: 180155

 Related applications: Anatomical Pathology

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top


CD30 is found on Reed-Sternberg cells and mononuclear Hodgkin's cells. Also reacts with anaplastic large cell lymphoma. May be useful in studying lymphomatoid papulosis. Positive tissue: Lymph node/Tonsil.

Clone/PAD: Ber-H2

Immunogen: CO cell line cells

Specificity: Ki-1 (CD30)

Reactivity: Human

Applications: Immunohistochemistry (FFPE)

Pretreatment: Epitope Retrieval: Required (EDTA, pH 8), Enzyme Digestion: Not required

Expected Staining Pattern: Primarily membrane with limited cytoplasmic
For In Vitro Diagnostic Use.


Reactivity: Human
Validated Application: Immunohistochemistry
Host Species: Mouse
SubClass (Isotype): IgG1
Clonality: Monoclonal
Target Gene: CD30
Gene ID: 943
Label or Dye: Unconjugated
Purification: Affinity Purified
Product Size: 1 mL
Research Category: Signal Transduction
Shipping Condition: Wet Ice

Contents & storage

Store at 2–8°C.


Manuals & protocols