Multi-Drug Resistance 1 (MDR1) Mouse Monoclonal Antibody (clone JSB-1), Concentrate

Catalog number: 180160Z

 Related applications: Anatomical Pathology

Error loading your content!

  Catalog number
Select a plan
Unit size
Price ({{currency}}) Your price ({{currency}}) Availability Qty
{{product.sku}} {{product.sku}}
also known as {{product.formattedSku}} 
{{subscriptionDetails.selectedRatePlan.billingPeriod}} {{subscriptionDetails.selectedRatePlan.billingPeriod}} {{product.availability.message}}
Pro add-ons

Your on-site stock

›› {{supplyCenter.scName}}({{scProduct.stockOnHand}} In stock)
›› {{supplyCenter.scName}}(Out of stock)
›› {{supplyCenter.scName}}
This item is not currently available on-site. Depending on your Supply Center settings you may be able to add the item to cart above else use the Order Non-Stocked Items' tab on the Supply Center home page.
Back to top

Description

This monoclonal antibody reacts with MDR. Elevated levels of p-glycoprotein have been reported in multidrug-resistant cell lines and in colon, endometrial, ovarian, and breast tumors, as well as in sarcomas and leukemias/lymphomas. Clone JSB-1 identifies an independent, strongly conserved cytoplasmic epitope present on the p-glycoprotein molecule, and does not cross-react with MDR3 p-glycoprotein. Positive tissue: Colon/liver.

Clone/PAD: JSB-1

Immunogen: Multi-drug resistant Chinese hamster cell line (CHrC5)

Specificity: Reacts with an independent, strongly conserved cytoplasmic epitope present on the p-glycoprotein molecule, and does not cross-react with MDR3 p-glycoprotein.

Reactivity: Human

Applications: Immunohistochemistry (FFPE)

Pretreatment: Epitope Retrieval: Required (Citrate buffer), Enzyme Digestion: Not required

Expected Staining Pattern: Membrane
For In Vitro Diagnostic Use.

Specifications

Reactivity: Human
Validated Application: Immunohistochemistry (FFPE)
Host Species: Mouse
SubClass (Isotype): IgG1
Clonality: Monoclonal
Target Gene: Multi-Drug Resistance 1 (MDR, p-Glycoprotein)
Gene ID: 4363, 5243, 5244
Label or Dye: Unconjugated
Dilution: Concentrate Antibody
Product Size: 1 mL
Shipping Condition: Wet Ice

Contents & storage

Store in refrigerator (2–8°C).

Documents

Manuals & protocols